missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli Chaperone Protein FIMC Protein
A cDNA sequence encoding the Chaperone Protein FIMC was constructed and used to recombinantly synthesize the protein.
1615.00 SEK - 46290.00 SEK
Specifications
Name | Chaperone Protein FIMC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | E. coli |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15997148
|
enQuireBio™
QP11402-5ug |
5 μg |
1615.00 SEK
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15987148
|
enQuireBio™
QP11402-25ug |
25 μg |
2515.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15977148
|
enQuireBio™
QP11402-1mg |
1 mg |
46290.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Chaperone Protein FIMC Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
The FIMC solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGVALGATRV IYPAGQKQEQ LAVTNNDENS TYLIQSWVEN ADGVKDGRFI VTPPLFAMKG KKENTLRILD ATNNQLPQDR ESLFWMNVKA IPSMDKSKLT ENTLQLAIIS RIKLYYRPAK LALPPDQAAE KLRFRRSANS LTLINPTPYY LTVTELNAGT RVLENALVPP MGESTVKLPS DAGSNITYRT INDYGALTPK MTGVME | |
Greater than 95.0% as determined by SDS-PAGE. |