missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cystathionine beta-lyase MetC Protein
A DNA sequence encoding the Escherichia coli (strain K12) Cystathionine beta-lyase MetC, was expressed in the hosts and tags indicated.
3070.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_417481.1 |
---|---|
Gene ID (Entrez) | 946240 |
Name | Cystathionine beta-lyase MetC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016541
|
enQuireBio™
QP8801-ec-10ug |
10 μg | N/A | ||||||
16056541
|
enQuireBio™
QP8801-ec-50ug |
50 μg | N/A | ||||||
16006541
|
enQuireBio™
QP8801-ec-100ug |
100 μg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16036541
|
enQuireBio™
QP8801-ec-200ug |
200 μg | N/A | ||||||
16046541
|
enQuireBio™
QP8801-ec-500ug |
500 μg | N/A | ||||||
16026541
|
enQuireBio™
QP8801-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_417481.1 | |
Cystathionine beta-lyase MetC Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
946240 | |
Research Use Only | |
metC | |
Recombinant Protein | |
E. coli | |
MADKKLDTQL VNAGRSKKYT LGAVNSVIQR ASSLVFDSVE AKKHATRNRA NGELFYGRRG TLTHFSLQQA MCELEGGAGC VLFPCGAAAV ANSILAFIEQ GDHVLMTNTA YEPSQDFCSK ILSKLGVTTS WFDPLIGADI VKHLQPNTKI VFLESPGSIT MEVHDVPAIV AAVRSVVPDA IIMIDNTWAA GVLFKALDFG IDVSIQAATK YLVGHSDAMI GTAVCNARCW EQLRENAYLM GQMVDADTAY ITSRGLRTLG VRLRQHHESS LKVAEWLAEH PQVARVNHPA LPGSKGHEFW KRDFTGSSGL FSFVLKKKLN NEELANYLDN FSLFSMAYSW GGYESLILAN QPEHIAAIRP QGEIDFSGTL IRLHIGLEDV DDLIADLDAG FA | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title