missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cytosine deaminase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Cytosine deaminase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7203-ye-1mg

 View more versions of this product

Product Code. 16057690

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_414871.1
Cytosine deaminase Protein
Research Use Only
codA
Recombinant Protein
Yeast
SNNALQTIIN ARLPGEEGLW QIHLQDGKIS AIDAQSGVMP ITENSLDAEQ GLVIPPFVEP HIHLDTTQTA GQPNWNQSGT LFEGIERWAE RKALLTHDDV KQRAWQTLKW QIANGIQHVR THVDVSDATL TALKAMLEVK QEVAPWIDLQ IVAFPQEGIL SYPNGEALLE EALRLGADVV GAIPHFEFTR EYGVESLHKT FALAQKYDRL IDVHCDEIDD EQSRFVETVA ALAHHEGMGA RVTASHTTAM HSYNGAYTSR LFRLLKMSGI NFVANPLVNI HLQGRFDTYP KRRGITRVKE MLESGINVCF GHDDVFDPWY PLGTANMLQV LHMGLHVCQL MGYGQINDGL NLITHHSART LNLQDYGIAA GNSANLIILP AENGFDALRR QVPVRYSVRG GKVIASTQPA QTTVYLEQPE AIDYKR
Greater than 90% as determined by SDS-PAGE.
944996
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.