missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glutaminase 1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glutaminase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6951-500ug
This item is not returnable.
View return policy
Specifications
NP_415018.1 | |
Glutaminase 1 Protein | |
Research Use Only | |
glsA1 | |
Recombinant Protein | |
E. coli | |
MLDANKLQQA VDQAYTQFHS LNGGQNADYI PFLANVPGQL AAVAIVTCDG NVYSAGDSDY RFALESISKV CTLALALEDV GPQAVQDKIG ADPTGLPFNS VIALELHGGK PLSPLVNAGA IATTSLINAE NVEQRWQRIL HIQQQLAGEQ VALSDEVNQS EQTTNFHNRA IAWLLYSAGY LYCDAMEACD VYTRQCSTLL NTIELATLGA TLAAGGVNPL THKRVLQADN VPYILAEMMM EGLYGRSGDW AYRVGLPGKS GVGGGILAVV PGVMGIAAFS PPLDEDGNSV RGQKMVASVA KQLGYNVFKG | |
Greater than 90% as determined by SDS-PAGE. |
946187 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction