Learn More
enQuireBio™ Recombinant E. coli Heat-labile enterotoxin B chain Protein
A DNA sequence encoding the Escherichia coli (strain K12) Heat-labile enterotoxin B chain, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7016-50ug
 View more versions of this product
Product Code. 16017060
  • 7415.00 SEK / 50µg
Explore more special offers
This item is not returnable. View return policy
Specifications
WP_024167713.1
50 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Heat-labile enterotoxin B chain Protein
Research Use Only
eltB
Recombinant Protein
E. coli
APQSITELCS EYHNTQIYTI NDKILSYTES MAGKREMVII TFKSGATFQV EVPGSQHIDS QKKAIERMKD TLRITYLTET KIDKLCVWNN KTPNSIAAIS MEN
Greater than 90% as determined by SDS-PAGE.