missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Malate synthase G Protein

A DNA sequence encoding the Escherichia coli (strain K12) Malate synthase G, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7126-100ug

 View more versions of this product

Product Code. 16017450

  • 12630.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_417450.1
Malate synthase G Protein
Research Use Only
glcB
Recombinant Protein
E. coli
SQTITQSRLR IDANFKRFVD EEVLPGTGLD AAAFWRNFDE IVHDLAPENR QLLAERDRIQ AALDEWHRSN PGPVKDKAAY KSFLRELGYL VPQPERVTVE TTGIDSEITS QAGPQLVVPA MNARYALNAA NARWGSLYDA LYGSDIIPQE GAMVSGYDPQ RGEQVIAWVR RFLDESLPLE NGSYQDVVAF KVVDKQLRIQ LKNGKETTLR TPAQFVGYRG DAAAPTCILL KNNGLHIELQ IDANGRIGKD DPAHINDVIV EAAISTILDC EDSVAAVDAE DKILLYRNLL GLMQGTLQEK MEKNGRQIVR KLNDDRHYTA ADGSEISLHG RSLLFIRNVG HLMTIPVIWD SEGNEIPEGI LDGVMTGAIA LYDLKVQKNS RTGSVYIVKP KMHGPQEVAF ANKLFTRIET MLGMAPNTLK MGIMDEERRT SLNLRSCIAQ ARNRVAFINT GFLDRTGDEM HSVMEAGPML RKNQMKSTPW IKAYERNNVL SGLFCGLRGK AQIGKGMWAM PDLMADMYSQ KGDQLRAGAN TAWVPSPTAA TLHALHYHQT NVQSVQANIA QTEFNAEFEP LLDDLLTIPV AENANWSAQE IQQELDNNVQ GILGYVVRWV EQGIGCSKVP DIHNVALMED RATLRISSQH IANWLRHGIL TKEQVQASLE NMAKVVDQQN AGDPAYRPMA GNFANSCAFK AASDLIFLGV KQPNGYTEPL LHAWRLREKE SH
Greater than 90% as determined by SDS-PAGE.
948857
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers