missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Protein FimH Protein
A DNA sequence encoding the Escherichia coli (strain K12) Protein FimH, was expressed in the hosts and tags indicated.
3070.00 SEK - 40175.00 SEK
Specifications
Accession Number | NP_418740.1 |
---|---|
Gene ID (Entrez) | 948847 |
Name | Protein FimH Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16018530
|
enQuireBio™
QP7372-ec-10ug |
10 μg | N/A | ||||||
16058530
|
enQuireBio™
QP7372-ec-50ug |
50 μg | N/A | ||||||
16008530
|
enQuireBio™
QP7372-ec-100ug |
100 μg |
11590.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16038530
|
enQuireBio™
QP7372-ec-200ug |
200 μg | N/A | ||||||
16048530
|
enQuireBio™
QP7372-ec-500ug |
500 μg | N/A | ||||||
16028530
|
enQuireBio™
QP7372-ec-1mg |
1 mg |
40175.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_418740.1 | |
Protein FimH Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-GST | |
Tris-based buffer, 50% glycerol |
948847 | |
Research Use Only | |
fimH | |
Recombinant Protein | |
E. coli | |
FACKTANGTA IPIGGGSANV YVNLAPVVNV GQNLVVDLST QIFCHNDYPE TITDYVTLQR GSAYGGVLSN FSGTVKYSGS SYPFPTTSET PRVVYNSRTD KPWPVALYLT PVSSAGGVAI KAGSLIAVLI LRQTNNYNSD DFQFVWNIYA NNDVVVPTGG CDVSARDVTV TLPDYPGSVP IPLTVYCAKS QNLGYYLSGT TADAGNSIFT NTASFSPAQG VGVQLTRNGT IIPANNTVSL GAVGTSAVSL GLTANYARTG GQVTAGNVQS IIGVTFVYQ | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title