missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Apoptosis regulator BHRF1 Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Apoptosis regulator BHRF1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7005-500ug
Additional Details : Weight : 0.01000kg
Specifications
YP_401646.1 | |
Apoptosis regulator BHRF1 Protein | |
Research Use Only | |
BHRF1 | |
Recombinant Protein | |
E. coli | |
MAYSTREILL ALCIRDSRVH GNGTLHPVLE LAARETPLRL SPEDTVVLRY HVLLEEIIER NSETFTETWN RFITHTEHVD LDFNSVFLEI FHRGDPSLGR ALAWMAWCMH ACRTLCCNQS TPYYVVDLSV RGMLEASEGL DG | |
Greater than 80% as determined by SDS-PAGE. |
3783706 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His | |
Tris-based buffer, 50% glycerol |