missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Apoptosis regulator BHRF1 Protein

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Apoptosis regulator BHRF1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7005-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16087000

  • 12125.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

YP_401646.1
Apoptosis regulator BHRF1 Protein
Research Use Only
BHRF1
Recombinant Protein
E. coli
MAYSTREILL ALCIRDSRVH GNGTLHPVLE LAARETPLRL SPEDTVVLRY HVLLEEIIER NSETFTETWN RFITHTEHVD LDFNSVFLEI FHRGDPSLGR ALAWMAWCMH ACRTLCCNQS TPYYVVDLSV RGMLEASEGL DG
Greater than 80% as determined by SDS-PAGE.
3783706
100 μg
Not determined.
Partial (see sequence information for more details).
Epstein-Barr
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers