missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein GP350 Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein GP350, was expressed in the hosts and tags indicated.
4340.00 SEK - 38570.00 SEK
Specifications
Accession Number | YP_401667.1 |
---|---|
Gene ID (Entrez) | 3783713 |
Name | Envelope glycoprotein GP350 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16018380
|
enQuireBio™
QP7351-ec-10ug |
10 μg |
4550.00 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16058380
|
enQuireBio™
QP7351-ec-50ug |
50 μg | |||||||
16008380
|
enQuireBio™
QP7351-ec-100ug |
100 μg |
11125.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16038380
|
enQuireBio™
QP7351-ec-200ug |
200 μg | |||||||
16048380
|
enQuireBio™
QP7351-ec-500ug |
500 μg | |||||||
16028380
|
enQuireBio™
QP7351-ec-1mg |
1 mg |
38570.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
YP_401667.1 | |
Envelope glycoprotein GP350 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3783713 | |
Research Use Only | |
BLLF1 | |
Recombinant Protein | |
E. coli | |
MEAALLVCQY TIQSLIHLTG EDPGFFNVEI PEFPFYPTCN VCTADVNVTI NFDVGGKKHQ LDLDFGQLTP HTKAVYQPRG AFGGSENATN LFLLELLGAG ELALTMRSKK LPINVTTGEE QQVSLESVDV YFQDVFGTMW CHHAEMQNPV YLIPETVPYI KWDNCNSTNI TAVVRAQGLD VTLPLSLPTS AQDSNFSVKT EMLGNEIDIE CIMEDGEISQ VLPGDNKFNI TCSGYESHVP SGGILTSTSP VATPIPGTGY AYSLRLTPRP VSRFLGNNSI LYVFYSGNGP KASGGDYCIQ SNIVFSDEIP ASQDMPTNTT DITYVGDNAT YSVPMVTSED ANSPNVTVTA FWAWPNNTET DFKCKWTLTS GTPSGCENIS GAFASNRTFD ITVSGLGTAP KTLIITRTAT NATTTTHKVI FSKAPESTTT SPTLNTTGFA DPNTTTGLPS STHVPTNLTA PASTGPTVST ADVTSPTPAG TTSGASPVTP SPS | |
Greater than 80% as determined by SDS-PAGE. |