Learn More
enQuireBio™ Recombinant hepatitis Non-structural polyprotein pORF1 Protein
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Non-structural polyprotein pORF1, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7125-50ug
 View more versions of this product
Product Code. 16007450
  • 7415.00 SEK / 50µg
Explore more special offers
This item is not returnable. View return policy
Specifications
hepatitis Non-structural polyprotein pORF1 Protein
Research Use Only
ORF1
Recombinant Protein
E. coli
EVFWNHPIQR VIHNELELYC RARSGRCLEI GAHPRSINDN PNVVHRCFLR PAGRDVQRWY TAPTRGPAAN CRRSALRGLP AADRTYCFDG FSGCNFPAET GIALYSLHDM SPSDVAEAMF RHGMTRLYAA LHLPPEVLLP PGTYRTASYL LIHDGRRVVV TYEGDTSAGY NHDVSNLRSW I
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Full Length
Hepatitis
His
Tris-based buffer, 50% glycerol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant hepatitis Non-structural polyprotein pORF1 Protein > 50μg