missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant hepatitis Non-structural polyprotein pORF1 Protein
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Non-structural polyprotein pORF1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7125-200ug
This item is not returnable.
View return policy
Specifications
hepatitis Non-structural polyprotein pORF1 Protein | |
Research Use Only | |
ORF1 | |
Recombinant Protein | |
E. coli | |
EVFWNHPIQR VIHNELELYC RARSGRCLEI GAHPRSINDN PNVVHRCFLR PAGRDVQRWY TAPTRGPAAN CRRSALRGLP AADRTYCFDG FSGCNFPAET GIALYSLHDM SPSDVAEAMF RHGMTRLYAA LHLPPEVLLP PGTYRTASYL LIHDGRRVVV TYEGDTSAGY NHDVSNLRSW I | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction