missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Serum amyloid A Protein
A DNA sequence encoding the Equus caballus (Horse) Serum amyloid A protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6647-ye-100ug
This item is not returnable.
View return policy
Specifications
Serum amyloid A Protein | |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
Yeast | |
LLSFLGEAAR GTWDMIRAYN DMREANYIGA DKYFHARGNY DAAKRGPGGA WAAKVISDAR ENFQRFTDRF SFGGSGRGAE DSRADQAANE WGRSGKDPNH FRPHGLPDKY | |
Greater than 80% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Horse | |
His | |
Tris-based buffer, 50% glycerol |