missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S11 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S11, was expressed in the hosts and tags indicated.
1640.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_001006.1 |
---|---|
Gene ID (Entrez) | 6205 |
Name | 40S ribosomal protein S11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16014371
|
enQuireBio™
QP8389-ec-10ug |
10 μg | N/A | ||||||
16054371
|
enQuireBio™
QP8389-ec-50ug |
50 μg | N/A | ||||||
16004371
|
enQuireBio™
QP8389-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16034371
|
enQuireBio™
QP8389-ec-200ug |
200 μg | N/A | ||||||
16044371
|
enQuireBio™
QP8389-ec-500ug |
500 μg | N/A | ||||||
16024371
|
enQuireBio™
QP8389-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001006.1 | |
40S ribosomal protein S11 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6205 | |
Research Use Only | |
RPS11 | |
Recombinant Protein | |
E. coli | |
ADIQTERAYQ KQPTIFQNKK RVLLGETGKE KLPRYYKNIG LGFKTPKEAI EGTYIDKKCP FTGNVSIRGR ILSGVVTKMK MQRTIVIRRD YLHYIRKYNR FEKRHKNMSV HLSPCFRDVQ IGDIVTVGEC RPLSKTVRFN VLKVTKAAGT KKQFQKF | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title