missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANTXR1 Protein
A DNA sequence encoding the Homo sapiens (Human) ANTXR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8977-ye-100ug
This item is not returnable.
View return policy
Specifications
NP_060623.2 | |
ANTXR1 Protein | |
Research Use Only | |
Antxr1 | |
Recombinant Protein | |
Yeast | |
EDGGPACYGG FDLYFILDKS GSVLHHWNEI YYFVEQLAHK FISPQLRMSF IVFSTRGTTL MKLTEDREQI RQGLEELQKV LPGGDTYMHE GFERASEQIY YENRQGYRTA SVIIALTDGE LHEDLFFYSE REANRSRDLG AIVYCVGVKD FNETQLARIA DSKDHVFPVN DGFQALQGII HSILKKSCIE ILAAEPSTIC AGESFQVVVR GNGFRHARNV DRVLCSFKIN DSVTLNEKPF SVEDTYLLCP APILKEVGMK AALQVSMNDG LSFISSSVII TTTHCSDGS | |
Greater than 90% as determined by SDS-PAGE. |
84168 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |