missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASH2L Protein
A DNA sequence encoding the Homo sapiens (Human) ASH2L, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001098684.1 |
---|---|
Gene ID (Entrez) | 9070 |
Name | ASH2L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013131
|
enQuireBio™
QP8184-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16033131
|
enQuireBio™
QP8184-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023131
|
enQuireBio™
QP8184-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043131
|
enQuireBio™
QP8184-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053131
|
enQuireBio™
QP8184-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063131
|
enQuireBio™
QP8184-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001098684.1 | |
ASH2L Protein | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9070 | |
Research Use Only | |
ASH2L | |
Recombinant Protein | |
E. coli | |
MDTQAGSVDE ENGRQLGEVE LQCGICTKWF TADTFGIDTS SCLPFMTNYS FHCNVCHHSG NTYFLRKQAN LKEMCLSALA NLTWQSRTQD EHPKTMFSKD KDIIPFIDKY WECMTTRQRP GKMTWPNNIV KTMSKERDVF LVKEHPDPGS KDPEEDYPKF GLLDQDLSNI GPAYDNQKQS SAVSTSGNLN GGIAAGSSGK GRGAKRKQQD GGTTGTTKKA RSDPLFSAQR LPPHGYPLEH PFNKDGYRYI LAEPDPHAPD PEKLELDCWA GKPIPGDLYR ACLYERVLLA LHDRAPQLKI SDDRLTVVGE KGYSMVRASH GVRKGAWYFE ITVDEMPPDT AARLGWSQPL GNLQAPLGYD KFSYSWRSKK GTKFHQSIGK HYSSGYGQGD VLGFYINLPE DTETAKSLPD TYKDKALIKF KSYLYFEEKD FVDKAEKSLK QTPHSEIIFY KNGVNQGVAY KDIFEGVYFP AISLYKSCTV SINFGPCFKY PPKDLTYRPM SDMGWGAVVE HTLADVLYHV ETEVDGRRSP PWEP | |
Greater than 90% as determined by SDS-PAGE. |