missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCHE / Butyrylcholinesterase Protein

A DNA sequence encoding the Homo sapiens (Human) BCHE / Butyrylcholinesterase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9007-ye-10ug

 View more versions of this product

Product Code. 16087431

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000046.1
BCHE / Butyrylcholinesterase Protein
Research Use Only
BCHE
Recombinant Protein
Yeast
EDDIIIATKN GKVRGMNLTV FGGTVTAFLG IPYAQPPLGR LRFKKPQSLT KWSDIWNATK YANSCCQNID QSFPGFHGSE MWNPNTDLSE DCLYLNVWIP APKPKNATVL IWIYGGGFQT GTSSLHVYDG KFLARVERVI VVSMNYRVGA LGFLALPGNP EAPGNMGLFD QQLALQWVQK NIAAFGGNPK SVTLFGESAG AASVSLHLLS PGSHSLFTRA ILQSGSFNAP WAVTSLYEAR NRTLNLAKLT GCSRENETEI IKCLRNKDPQ EILLNEAFVV PYGTPLSVNF GPTVDGDFLT DMPDILLELG QFKKTQILVG VNKDEGTAFL VYGAPGFSKD NNSIITRKEF QEGLKIFFPG VSEFGKESIL FHYTDWVDDQ RPENYREALG DVVGDYNFIC PALEFTKKFS EWGNNAFFYY FEHRSSKLPW PEWMGVMHGY EIEFVFGLPL ERRDNYTKAE EILSRSIVKR WANFAKYGNP NETQNNSTSW PVFKSTEQKY LTLNTESTRI MTKLRAQQCR FWTSFFPKVL EMTGNIDEAE WEWKAGFHRW NNYMMDWKNQ FNDYTSKKES CVGL
Greater than 90% as determined by SDS-PAGE.
590
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers