missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCHE / Butyrylcholinesterase Protein
A DNA sequence encoding the Homo sapiens (Human) BCHE / Butyrylcholinesterase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9007-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_000046.1 | |
BCHE / Butyrylcholinesterase Protein | |
Research Use Only | |
BCHE | |
Recombinant Protein | |
Yeast | |
EDDIIIATKN GKVRGMNLTV FGGTVTAFLG IPYAQPPLGR LRFKKPQSLT KWSDIWNATK YANSCCQNID QSFPGFHGSE MWNPNTDLSE DCLYLNVWIP APKPKNATVL IWIYGGGFQT GTSSLHVYDG KFLARVERVI VVSMNYRVGA LGFLALPGNP EAPGNMGLFD QQLALQWVQK NIAAFGGNPK SVTLFGESAG AASVSLHLLS PGSHSLFTRA ILQSGSFNAP WAVTSLYEAR NRTLNLAKLT GCSRENETEI IKCLRNKDPQ EILLNEAFVV PYGTPLSVNF GPTVDGDFLT DMPDILLELG QFKKTQILVG VNKDEGTAFL VYGAPGFSKD NNSIITRKEF QEGLKIFFPG VSEFGKESIL FHYTDWVDDQ RPENYREALG DVVGDYNFIC PALEFTKKFS EWGNNAFFYY FEHRSSKLPW PEWMGVMHGY EIEFVFGLPL ERRDNYTKAE EILSRSIVKR WANFAKYGNP NETQNNSTSW PVFKSTEQKY LTLNTESTRI MTKLRAQQCR FWTSFFPKVL EMTGNIDEAE WEWKAGFHRW NNYMMDWKNQ FNDYTSKKES CVGL | |
Greater than 90% as determined by SDS-PAGE. |
590 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |