missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BMP-7 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

6640.00 SEK - 53025.00 SEK

Specifications

Gene ID (Entrez) 655
Name BMP-7
Source Chinese Hamster Ovary cell line,CHO-derived human BMP-7 protein Ser293-His431
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18311438
View Documents
R&D Systems™
354-BP-010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
6640.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
18106790
View Documents
R&D Systems™
354-BP-010/CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
6640.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
18654749
View Documents
R&D Systems™
354-BP-500/CF
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
53025.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human BMP-7 Protein is derived from CHO. The Recombinant Human BMP-7 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

655
Chinese Hamster Ovary cell line,CHO-derived human BMP-7 protein Ser293-His431
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
BMP-7
Disulfide-linked homodimer
Recombinant
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
R&D Systems™ Recombinant Human BMP-7 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.