missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1D Protein
A DNA sequence encoding the Homo sapiens (Human) C1D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP938-200ug
This item is not returnable.
View return policy
Specifications
NP_001177192.1 | |
C1D Protein | |
Research Use Only | |
C1D | |
Recombinant Protein | |
E. coli | |
MAGEEINEDY PVEIHEYLSA FENSIGAVDE MLKTMMSVSR NELLQKLDPL EQAKVDLVSA YTLNSMFWVY LATQGVNPKE HPVKQELERI RVYMNRVKEI TDKKKAGKLD RGAASRFVKN ALWEPKSKNA SKVAN | |
Greater than 90% as determined by SDS-PAGE. |
10438 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction