missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CPVL / Carboxypeptidase Protein

A DNA sequence encoding the Homo sapiens (Human) CPVL / Carboxypeptidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8142-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092871

  • 5980.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001334981.1
CPVL / Carboxypeptidase Protein
Research Use Only
CPVL
Recombinant Protein
E. coli
LFRSLYRSVS MPPKGDSGQP LFLTPYIEAG KIQKGRELSL VGPFPGLNMK SYAGFLTVNK TYNSNLFFWF FPAQIQPEDA PVVLWLQGGP GGSSMFGLFV EHGPYVVTSN MTLRDRDFPW TTTLSMLYID NPVGTGFSFT DDTHGYAVNE DDVARDLYSA LIQFFQIFPE YKNNDFYVTG ESYAGKYVPA IAHLIHSLNP VREVKINLNG IAIGDGYSDP ESIIGGYAEF LYQIGLLDEK QKKYFQKQCH ECIEHIRKQN WFEAFEILDK LLDGDLTSDP SYFQNVTGCS NYYNFLRCTE PEDQLYYVKF LSLPEVRQAI HVGNQTFNDG TIVEKYLRED TVQSVKPWLT EIMNNYKVLI YNGQLDIIVA AALTERSLMG MDWKGSQEYK KAEKKVWKIF KSDSEVAGYI RQAGDFHQVI IRGGGHILPY DQPLRAFDMI NRFIYGKGWD PYVG
Greater than 90% as determined by SDS-PAGE.
54504
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers