missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDB1- and CUL4-associated factor 7 Protein
A DNA sequence encoding the Homo sapiens (Human) DDB1- and CUL4-associated factor 7, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_005819.3 |
---|---|
Gene ID (Entrez) | 10238 |
Name | DDB1- and CUL4-associated factor 7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016550
|
enQuireBio™
QP6885-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036550
|
enQuireBio™
QP6885-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026550
|
enQuireBio™
QP6885-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046550
|
enQuireBio™
QP6885-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056550
|
enQuireBio™
QP6885-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066550
|
enQuireBio™
QP6885-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005819.3 | |
DDB1- and CUL4-associated factor 7 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10238 | |
Research Use Only | |
DCAF7 | |
Recombinant Protein | |
E. coli | |
MSLHGKRKEI YKYEAPWTVY AMNWSVRPDK RFRLALGSFV EEYNNKVQLV GLDEESSEFI CRNTFDHPYP TTKLMWIPDT KGVYPDLLAT SGDYLRVWRV GETETRLECL LNNNKNSDFC APLTSFDWNE VDPYLLGTSS IDTTCTIWGL ETGQVLGRVN LVSGHVKTQL IAHDKEVYDI AFSRAGGGRD MFASVGADGS VRMFDLRHLE HSTIIYEDPQ HHPLLRLCWN KQDPNYLATM AMDGMEVVIL DVRVPCTPVA RLNNHRACVN GIAWAPHSSC HICTAADDHQ ALIWDIQQMP RAIEDPILAY TAEGEINNVQ WASTQPDWIA ICYNNCLEIL RV | |
Greater than 90% as determined by SDS-PAGE. |