missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-16 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
2395.00 SEK
Specifications
Gene ID (Entrez) | 8823 |
---|---|
Molecular Weight (g/mol) | 23.7 kDa |
Name | FGF-16 |
Quantity | 25 μg |
Source | E. coli-derived human FGF-16 protein Ala2-Arg207 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18198710
|
R&D Systems™
1212-FG-025 |
Lyophilized from a 0.2 μm filtered solution in MOPS and Na 2 SO 4 with BSA as a carrier protein. |
2395.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18106910
|
R&D Systems™
1212-FG-025/CF |
Lyophilized from a 0.2 μm filtered solution in MOPS and Na 2 SO 4 . |
2395.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human FGF-16 Protein is derived from E. coli. The Recombinant Human FGF-16 Protein has been validated for the following applications: Bioactivity.Specifications
8823 | |
FGF-16 | |
E. coli-derived human FGF-16 protein Ala2-Arg207 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
23.7 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |