missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GALNT14 Protein
A DNA sequence encoding the Homo sapiens (Human) GALNT14, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_001240755.1 |
---|---|
Gene ID (Entrez) | 79623 |
Name | GALNT14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031291
|
enQuireBio™
QP7871-ec-10ug |
10 μg | N/A | ||||||
16071291
|
enQuireBio™
QP7871-ec-50ug |
50 μg | N/A | ||||||
16021291
|
enQuireBio™
QP7871-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16051291
|
enQuireBio™
QP7871-ec-200ug |
200 μg | N/A | ||||||
16061291
|
enQuireBio™
QP7871-ec-500ug |
500 μg | N/A | ||||||
16041291
|
enQuireBio™
QP7871-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001240755.1 | |
GALNT14 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79623 | |
Research Use Only | |
GALNT14 | |
Recombinant Protein | |
E. coli | |
MRRLTRRLVL PVFGVLWITV LLFFWVTKRK LEVPTGPEVQ TPKPSDADWD DLWDQFDERR YLNAKKWRVG DDPYKLYAFN QRESERISSN RAIPDTRHLR CTLLVYCTDL PPTSIIITFH NEARSTLLRT IRSVLNRTPT HLIREIILVD DFSNDPDDCK QLIKLPKVKC LRNNERQGLV RSRIRGADIA QGTTLTFLDS HCEVNRDWLQ PLLHRVKEDY TRVVCPVIDI INLDTFTYIE SASELRGGFD WSLHFQWEQL SPEQKARRLD PTEPIRTPII AGGLFVIDKA WFDYLGKYDM DMDIWGGENF EISFRVWMCG GSLEIVPCSR VGHVFRKKHP YVFPDGNANT YIKNTKRTAE VWMDEYKQYY YAARPFALER PFGNVESRLD LRKNLRCQSF KWYLENIYPE LSIPKESSIQ KGNIRQRQKC LESQRQNNQE TPNLKLSPCA KVKGEDAKSQ VWAFTYTQQI LQEELCLSVI TLFPGAPVVL VLCKNGDDRQ QWTKTGSHIE HIASHLCLDT DMFGDGTENG KEIVVNPCES SLMSQHWDMV SS | |
Greater than 90% as determined by SDS-PAGE. |