missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hexokinase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Hexokinase-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6164-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_000179.2 | |
Hexokinase-1 Protein | |
Research Use Only | |
HK1 | |
Recombinant Protein | |
Yeast | |
ELKDDQVKKI DKYLYAMRLS DETLIDIMTR FRKEMKNGLS RDFNPTATVK MLPTFVRSIP DGSEKGDFIA LDLGGSSFRI LRVQVNHEKN QNVHMESEVY DTPENIVHGS GSQLFDHVAE CLGDFMEKRK IKDKKLPVGF TFSFPCQQSK IDEAILITWT KRFKASGVEG ADVVKLLNKA IKKRGDYDAN IVAVVNDTVG TMMTCGYDDQ HCEVGLIIGT GTNACYMEEL RHIDLVEGDE GRMCINTEWG AFGDDGSLED IRTEFDREID RGSLNPGKQL FEKMVSGMYL GELVRLILVK MAKEGLLFEG RITPELLTRG KFNTSDVSAI EKNKEGLHNA KEILTRLGVE PSDDDCVSVQ HVCTIVSFRS ANLVAATLGA ILNRLRDNKG TPRLRTTVGV DGSLYKTHPQ YSRRFHKTLR RLVPDSDVRF LLSESGSGKG AAMVTAVAYR LAEQHRQIEE TLA | |
Greater than 90% as determined by SDS-PAGE. |
3098 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction