missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone deacetylase 6 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6154-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001308155.1 | |
Histone deacetylase 6 Protein | |
Research Use Only | |
HDAC6 | |
Recombinant Protein | |
E. coli | |
MMNHCNLWDS HHPEVPQRIL RIMCRLEELG LAGRCLTLTP RPATEAELLT CHSAEYVGHL RATEKMKTRE LHRESSNFDS IYICPSTFAC AQLATGAACR LVEAVLSGEV LNGAAVVRPP GHHAEQDAAC GFCFFNSVAV AARHAQTISG HALRILIVDW DVHHGNGTQH MFEDDPSVLY VSLHRYDHGT FFPMGDEGAS SQIGRAAGTG FTVNVAWNGP RMGDADYLAA WHRLVLPIAY EFNPELVLVS AGFDAARGDP LGGCQVSPEG YAHLTHLLMG LASGRIILIL EGGYNLTSIS ESMAACTRSL LGDPPPLLTL PRPPLSGALA SITETIQVHR RYWRSLRVMK VE | |
Greater than 80% as determined by SDS-PAGE. |
10013 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |