missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone deacetylase 6 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone deacetylase 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6154-ec-100ug

 View more versions of this product

Product Code. 16091200

  • 6235.00 SEK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001308155.1
Histone deacetylase 6 Protein
Research Use Only
HDAC6
Recombinant Protein
E. coli
MMNHCNLWDS HHPEVPQRIL RIMCRLEELG LAGRCLTLTP RPATEAELLT CHSAEYVGHL RATEKMKTRE LHRESSNFDS IYICPSTFAC AQLATGAACR LVEAVLSGEV LNGAAVVRPP GHHAEQDAAC GFCFFNSVAV AARHAQTISG HALRILIVDW DVHHGNGTQH MFEDDPSVLY VSLHRYDHGT FFPMGDEGAS SQIGRAAGTG FTVNVAWNGP RMGDADYLAA WHRLVLPIAY EFNPELVLVS AGFDAARGDP LGGCQVSPEG YAHLTHLLMG LASGRIILIL EGGYNLTSIS ESMAACTRSL LGDPPPLLTL PRPPLSGALA SITETIQVHR RYWRSLRVMK VE
Greater than 80% as determined by SDS-PAGE.
10013
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers