missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein NANOG Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein NANOG, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001284627.1 |
---|---|
Gene ID (Entrez) | 79923 |
Name | Homeobox protein NANOG Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053341
|
enQuireBio™
QP8217-ec-10ug |
10 μg | |||||||
16093341
|
enQuireBio™
QP8217-ec-50ug |
50 μg | |||||||
16043341
|
enQuireBio™
QP8217-ec-100ug |
100 μg |
5980.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16073341
|
enQuireBio™
QP8217-ec-200ug |
200 μg | |||||||
16083341
|
enQuireBio™
QP8217-ec-500ug |
500 μg | |||||||
16063341
|
enQuireBio™
QP8217-ec-1mg |
1 mg |
26990.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001284627.1 | |
Homeobox protein NANOG Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79923 | |
Research Use Only | |
NANOG | |
Recombinant Protein | |
E. coli | |
MSVDPACPQS LPCFEASDCK ESSPMPVICG PEENYPSLQM SSAEMPHTET VSPLPSSMDL LIQDSPDSST SPKGKQPTSA EKSVAKKEDK VPVKKQKTRT VFSSTQLCVL NDRFQRQKYL SLQQMQELSN ILNLSYKQVK TWFQNQRMKS KRWQKNNWPK NSNGVTQKAS APTYPSLYSS YHQGCLVNPT GNLPMWSNQT WNNSTWSNQT QNIQSWSNHS WNTQTWCTQS WNNQAWNSPF YNCGEESLQS CMQFQPNSPA SDLEAALEAA GEGLNVIQQT TRYFSTPQTM DLFLNYSMNM QPEDV | |
Greater than 90% as determined by SDS-PAGE. |