missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein

A DNA sequence encoding the Human IL-29 / Interleukin-29, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10410-ec-100ug

 View more versions of this product

Product Code. 15923768

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_742152.1
IL-29 / Interleukin-29 Protein
Research Use Only
IFNL1
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis is less than 5 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
282618
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GPVPTSKPTT TGKGCHIGRF KSLSPQELAS FKKARDALEE SLKLKNWSCS SPVFPGNWDL RLLQVRERPV ALEAELALTL KVLEAAAGPA LEDVLDQPLH TLHHILSQLQ ACIQPQPTAG PRPRGRLHHW LHRLQEAPKK ESAGCLEASV TFNLFRLLTR DLKYVADGNL CLRTSTHPES T
Greater than 97% as determined by SDS-PAGE and HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers