missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein
A DNA sequence encoding the Human IL-29 / Interleukin-29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10410-ec-250ug
This item is not returnable.
View return policy
Specifications
NP_742152.1 | |
IL-29 / Interleukin-29 Protein | |
Research Use Only | |
IFNL1 | |
The ED50 as determined by an anti-viral assay using human HepG2 cells infected with encephalomyocarditis is less than 5 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
282618 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPVPTSKPTT TGKGCHIGRF KSLSPQELAS FKKARDALEE SLKLKNWSCS SPVFPGNWDL RLLQVRERPV ALEAELALTL KVLEAAAGPA LEDVLDQPLH TLHHILSQLQ ACIQPQPTAG PRPRGRLHHW LHRLQEAPKK ESAGCLEASV TFNLFRLLTR DLKYVADGNL CLRTSTHPES T | |
Greater than 97% as determined by SDS-PAGE and HPLC. |