missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-3 / Interleukin-3 Protein

A DNA sequence encoding the Homo sapiens (Human) IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6220-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011690

  • 26990.00 SEK / 1mg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_000579.2
IL-3 / Interleukin-3 Protein
Research Use Only
IL3
Recombinant Protein
E. coli
APMTQTTPLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF
Greater than 90% as determined by SDS-PAGE.
3562
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers