missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL16 / Interleukin-16 Protein
A DNA sequence encoding the Human IL16 / Interleukin-16, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10303-ec-250ug
This item is not returnable.
View return policy
Specifications
NP_001165599.1 | |
IL16 / Interleukin-16 Protein | |
Research Use Only | |
IL16 | |
The biological activity determined by a chemotaxis bioassay using human CD4+ T Lymphocytes is in a concentration range of 1.0-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3603 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MPDLNSSTDS AASASAASDV SVESTAEATV CTVTLEKMSA GLGFSLEGGK GSLHGDKPLT INRIFKGAAS EQSETVQPGD EILQLGGTAM QGLTRFEAWN IIKALPDGPV TIVIRRKSLQ SKETTAAGDS | |
Greater than 95% as determined by SDS-PAGE and HPLC. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human IL16 / Interleukin-16 Protein > 250μg
Spot an opportunity for improvement?Share a Content Correction