missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Inactive phospholipase D5 Protein

A DNA sequence encoding the Homo sapiens (Human) Inactive phospholipase D5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7881-ec-200ug

 View more versions of this product

Product Code. 16041361

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001182740.1
Inactive phospholipase D5 Protein
Research Use Only
PLD5
Recombinant Protein
E. coli
MGEDEDGLSE KNCQNKCRIA LVENIPEGLN YSENAPFHLS LFQGWMNLLN MAKKSVDIVS SHWDLNHTHP SACQGQRLFE KLLQLTSQNI EIKLVSDVTA DSKVLEALKL KGAEVTYMNM TAYNKGRLQS SFWIVDKQHV YIGSAGLDWQ SLGQMKELGV IFYNCSCLVL DLQRIFALYS SLKFKSRVPQ TWSKRLYGVY DNEKKLQLQL NETKSQAFVS NSPKLFCPKN RSFDIDAIYS VIDDAKQYVY IAVMDYLPIS STSTKRTYWP DLDAKIREAL VLRSVRVRLL LSFWKETDPL TFNFISSLKA ICTEIANCSL KVKFFDLERE NACATKEQKN HTFPRLNRNK YMVTDGAAYI GNFDWVGNDF TQNAGTGLVI NQADVRNNRS IIKQLKDVFE RDWYSPYAKT LQPTKQPNCS SLFKLKPLSN KTATDDTGGK DPRNV
Greater than 90% as determined by SDS-PAGE.
200150
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers