missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Josephin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Josephin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7617-ec-200ug

 View more versions of this product

Product Code. 16069820

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_055691.1
Josephin-1 Protein
Research Use Only
JOSD1
Recombinant Protein
E. coli
MSCVPWKGDK AKSESLELPQ AAPPQIYHEK QRRELCALHA LNNVFQDSNA FTRDTLQEIF QRLSPNTMVT PHKKSMLGNG NYDVNVIMAA LQTKGYEAVW WDKRRDVGVI ALTNVMGFIM NLPSSLCWGP LKLPLKRQHW ICVREVGGAY YNLDSKLKMP EWIGGESELR KFLKHHLRGK NCELLLVVPE EVEAHQSWRT DV
Greater than 90% as determined by SDS-PAGE.
9929
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.