missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCND1 Protein
A DNA sequence encoding the Homo sapiens (Human) KCND1, was expressed in the hosts and tags indicated.
2005.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_004970.3 |
---|---|
Gene ID (Entrez) | 3750 |
Name | KCND1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16081940
|
enQuireBio™
QP6253-ye-10ug |
10 μg | |||||||
16021950
|
enQuireBio™
QP6253-ye-50ug |
50 μg | |||||||
16071940
|
enQuireBio™
QP6253-ye-100ug |
100 μg |
7265.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16001950
|
enQuireBio™
QP6253-ye-200ug |
200 μg | |||||||
16011950
|
enQuireBio™
QP6253-ye-500ug |
500 μg |
19700.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16091940
|
enQuireBio™
QP6253-ye-1mg |
1 mg | |||||||
Specifications
NP_004970.3 | |
KCND1 Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3750 | |
Research Use Only | |
KCND1 | |
Recombinant Protein | |
Yeast | |
NFSRIYHQNQ RADKRRAQQK VRLARIRLAK SGTTNAFLQY KQNGGLEDSG SGEEQALCVR NRSAFEQQHH HLLHCLEKTT CHEFTDELTF SEALGAVSPG GRTSRSTSVS SQPVGPGSLL SSCCPRRAKR RAIRLANSTA SVSRGSMQEL DMLAGLRRSH APQSRSSLNA KPHDSLDLNC DSRDFVAAII SIPTPPANTP DESQPSSPGG GGRAGSTLRN SSLGTPCLFP ETVKISSL | |
Greater than 90% as determined by SDS-PAGE. |