missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leucine-rich repeat-containing protein 40 Protein
A DNA sequence encoding the Homo sapiens (Human) Leucine-rich repeat-containing protein 40, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8175-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_060238.3 | |
Leucine-rich repeat-containing protein 40 Protein | |
Research Use Only | |
LRRC40 | |
Recombinant Protein | |
E. coli | |
MSRLKRIAGQ DLRAGFKAGG RDCGTSVPQG LLKAARKSGQ LNLSGRNLSE VPQCVWRINV DIPEEANQNL SFGATERWWE QTDLTKLIIS NNKLQSLTDD LRLLPALTVL DIHDNQLTSL PSAIRELENL QKLNVSHNKL KILPEEITNL RNLKCLYLQH NELTCISEGF EQLSNLEDLD LSNNHLTTVP ASFSSLSSLV RLNLSSNELK SLPAEINRMK RLKHLDCNSN LLETIPPELA GMESLELLYL RRNKLRFLPE FPSCSLLKEL HVGENQIEML EAEHLKHLNS ILVLDLRDNK LKSVPDEIIL LRSLERLDLS NNDISSLPYS LGNLHLKFLA LEGNPLRTIR REIISKGTQE VLKYLRSKIK DDGPSQSESA TETAMTLPSE SRVNIHAIIT LKILDYSDKQ ATLIPDEVFD AVKSNIVTSI NFSKNQLCEI PKRMVELKEM VSDVDLSFNK LSFISLELCV LQKLTFLDLR NNFLNSLPEE MESLVRLQTI NLSFNRFKML PEVLYRIFTL ETILISNNQV GSVDPQKMKM MENLTTLDLQ NNDLLQIPPE LGNCVNLRTL LLDGNPFRVP RAAILMKGTA AILEYLRDRI PT | |
Greater than 90% as determined by SDS-PAGE. |
55631 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |