missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mannan-binding lectin serine protease 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Mannan-binding lectin serine protease 1, was expressed in the hosts and tags indicated.
2075.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_001027019.1 |
---|---|
Gene ID (Entrez) | 5648 |
Name | Mannan-binding lectin serine protease 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032650
|
enQuireBio™
QP6347-ye-10ug |
10 μg | |||||||
16072650
|
enQuireBio™
QP6347-ye-50ug |
50 μg | |||||||
16022650
|
enQuireBio™
QP6347-ye-100ug |
100 μg |
7265.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16052650
|
enQuireBio™
QP6347-ye-200ug |
200 μg | |||||||
16062650
|
enQuireBio™
QP6347-ye-500ug |
500 μg |
19700.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16042650
|
enQuireBio™
QP6347-ye-1mg |
1 mg | |||||||
Specifications
NP_001027019.1 | |
Mannan-binding lectin serine protease 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5648 | |
Research Use Only | |
MASP1 | |
Recombinant Protein | |
Yeast | |
HTVELNNMFG QIQSPGYPDS YPSDSEVTWN ITVPDGFRIK LYFMHFNLES SYLCEYDYVK VETEDQVLAT FCGRETTDTE QTPGQEVVLS PGSFMSITFR SDFSNEERFT GFDAHYMAVD VDECKEREDE ELSCDHYCHN YIGGYYCSCR FGYILHTDNR TCRVECSDNL FTQRTGVITS PDFPNPYPKS SECLYTIELE EGFMVNLQFE DIFDIEDHPE VPCPYDYIKI KVGPKVLGPF CGEKAPEPIS TQSHSVLILF HSDNSGENRG WRLSYRAAGN ECPELQPPVH GKIEPSQAKY FFKDQVLVSC DTGYKVLKDN VEMDTFQIEC LKDGTWSNKI PTCKIVDCRA PGELEHGLIT FSTRNNLTTY KSEIKYSCQE PYYKMLNNNT GIYTCSAQGV WMNKVLGRSL PTCLPVCGLP KFSRKLMARI FNGRPAQKGT TPWIAMLSHL NGQPFCGGSL LGSSWIVTAA HCLHQSLDPE DPTLRDSDLL SPSDFKIILG KHWRLRSDEN EQHLGVKHTT LHPQYDPNTF ENDVALVELL ESPVLNAFVM PICLPEGPQQ EGAMVIVSGW GKQFLQRFPE TLMEIEIPIV DHSTCQKAYA PLKKKVTRDM ICAGEKEGGK DACAGDSGGP MVTLNRERGQ WYLVGTVSWG DDCGKKDRYG VYSYIHHNKD WIQRVTGVRN | |
Greater than 90% as determined by SDS-PAGE. |