missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP2K3 Protein
A DNA sequence encoding the Homo sapiens (Human) MAP2K3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6336-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_001303261.1 | |
MAP2K3 Protein | |
Research Use Only | |
MAP2K3 | |
Recombinant Protein | |
E. coli | |
MESPASSQPA SMPQSKGKSK RKKDLRISCM SKPPAPNPTP PRNLDSRTFI TIGDRNFEVE ADDLVTISEL GRGAYGVVEK VRHAQSGTIM AVKRIRATVN SQEQKRLLMD LDINMRTVDC FYTVTFYGAL FREGDVWICM ELMDTSLDKF YRKVLDKNMT IPEDILGEIA VSIVRALEHL HSKLSVIHRD VKPSNVLINK EGHVKMCDFG ISGYLVDSVA KTMDAGCKPY MAPERINPEL NQKGYNVKSD VWSLGITMIE MAILRFPYES WGTPFQQLKQ VVEEPSPQLP ADRFSPEFVD FTAQCLRKNP AERMSYLELM EHPFFTLHKT KKTDIAAFVK EILGEDS | |
Greater than 90% as determined by SDS-PAGE. |
5606 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction