missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAP2K3 Protein

A DNA sequence encoding the Homo sapiens (Human) MAP2K3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6336-ec-10ug

 View more versions of this product

Product Code. 16022570

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001303261.1
MAP2K3 Protein
Research Use Only
MAP2K3
Recombinant Protein
E. coli
MESPASSQPA SMPQSKGKSK RKKDLRISCM SKPPAPNPTP PRNLDSRTFI TIGDRNFEVE ADDLVTISEL GRGAYGVVEK VRHAQSGTIM AVKRIRATVN SQEQKRLLMD LDINMRTVDC FYTVTFYGAL FREGDVWICM ELMDTSLDKF YRKVLDKNMT IPEDILGEIA VSIVRALEHL HSKLSVIHRD VKPSNVLINK EGHVKMCDFG ISGYLVDSVA KTMDAGCKPY MAPERINPEL NQKGYNVKSD VWSLGITMIE MAILRFPYES WGTPFQQLKQ VVEEPSPQLP ADRFSPEFVD FTAQCLRKNP AERMSYLELM EHPFFTLHKT KKTDIAAFVK EILGEDS
Greater than 90% as determined by SDS-PAGE.
5606
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.