missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MORF4 family-associated protein 1-like 1 Protein
A DNA sequence encoding the Homo sapiens (Human) MORF4 family-associated protein 1-like 1, was expressed in the hosts and tags indicated.
2075.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_982287.1 |
---|---|
Gene ID (Entrez) | 114932 |
Name | MORF4 family-associated protein 1-like 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16032890
|
enQuireBio™
QP6379-ye-10ug |
10 μg | N/A | ||||||
16072890
|
enQuireBio™
QP6379-ye-50ug |
50 μg | N/A | ||||||
16022890
|
enQuireBio™
QP6379-ye-100ug |
100 μg |
7570.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16052890
|
enQuireBio™
QP6379-ye-200ug |
200 μg | N/A | ||||||
16062890
|
enQuireBio™
QP6379-ye-500ug |
500 μg |
20530.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16042890
|
enQuireBio™
QP6379-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_982287.1 | |
MORF4 family-associated protein 1-like 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
114932 | |
Research Use Only | |
MRFAP1L1 | |
Recombinant Protein | |
Yeast | |
MRPLDIDEVE APEEVEVLEP EEDFEQFLLP VINEMREDIA SLIREHGRAY LRTRSKLWEM DNMLIQIKTQ VEASEESALN HVQHPSGEAD ERVSELCEKA EEKAKEIAKM AEMLVELVWR IERSESS | |
Greater than 90% as determined by SDS-PAGE. |