missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nicotinamide N-methyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Nicotinamide N-methyltransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6426-ec-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_006160.1 | |
Nicotinamide N-methyltransferase Protein | |
Research Use Only | |
NNMT | |
Recombinant Protein | |
E. coli | |
MESGFTSKDT YLSHFNPRDY LEKYYKFGSR HSAESQILKH LLKNLFKIFC LDGVKGDLLI DIGSGPTIYQ LLSACESFKE IVVTDYSDQN LQELEKWLKK EPEAFDWSPV VTYVCDLEGN RVKGPEKEEK LRQAVKQVLK CDVTQSQPLG AVPLPPADCV LSTLCLDAAC PDLPTYCRAL RNLGSLLKPG GFLVIMDALK SSYYMIGEQK FSSLPLGREA VEAAVKEAGY TIEWFEVISQ SYSSTMANNE GLFSLVARKL SRPL | |
Greater than 80% as determined by SDS-PAGE. |
4837 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |