missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMNAT1 / NMNAT Protein
A DNA sequence encoding the Homo sapiens (Human) NMNAT1 / NMNAT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8070-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001284707.1 | |
NMNAT1 / NMNAT Protein | |
Research Use Only | |
NMNAT1 | |
Recombinant Protein | |
E. coli | |
MENSEKTEVV LLACGSFNPI TNMHLRLFEL AKDYMNGTGR YTVVKGIISP VGDAYKKKGL IPAYHRVIMA ELATKNSKWV EVDTWESLQK EWKETLKVLR HHQEKLEASD CDHQQNSPTL ERPGRKRKWT ETQDSSQKKS LEPKTKAVPK VKLLCGADLL ESFAVPNLWK SEDITQIVAN YGLICVTRAG NDAQKFIYES DVLWKHRSNI HVVNEWIAND ISSTKIRRAL RRGQSIRYLV PDLVQEYIEK HNLYSSESED RNAGVILAPL QRNTAEAKT | |
Greater than 80% as determined by SDS-PAGE. |
64802 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |