missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human P4HB / ERBA2L Protein

A DNA sequence encoding the Homo sapiens (Human) P4HB / ERBA2L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6460-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033460

  • 31280.00 SEK / 1mg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_000909.2
P4HB / ERBA2L Protein
Research Use Only
P4hb
Recombinant Protein
Yeast
DAPEEEDHVL VLRKSNFAEA LAAHKYLLVE FYAPWCGHCK ALAPEYAKAA GKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFRN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLPDGAAAE SLVESSEVAV IGFFKDVESD SAKQFLQAAE AIDDIPFGIT SNSDVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKENLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLSNFKT AAESFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESEELTAE RITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EDVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIVIAKMDS TANEVEAVKV HSFPTLKFFP ASADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDLEDL EEAEEPDMEE DDDQKAVKDE L
Greater than 90% as determined by SDS-PAGE.
5034
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers