missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Plastin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Plastin-2, was expressed in the hosts and tags indicated.

1640.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_002289.2
Gene ID (Entrez) 3936
Name Plastin-2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16022210
View Documents
enQuireBio™
QP6289-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16062210
View Documents
enQuireBio™
QP6289-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16012210
View Documents
enQuireBio™
QP6289-ec-100ug
100 μg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16042210
View Documents
enQuireBio™
QP6289-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16052210
View Documents
enQuireBio™
QP6289-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16032210
View Documents
enQuireBio™
QP6289-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_002289.2
Plastin-2 Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3936
Research Use Only
LCP1
Recombinant Protein
E. coli
ARGSVSDEEM MELREAFAKV DTDGNGYISF NELNDLFKAA CLPLPGYRVR EITENLMATG DLDQDGRISF DEFIKIFHGL KSTDVAKTFR KAINKKEGIC AIGGTSEQSS VGTQHSYSEE EKYAFVNWIN KALENDPDCR HVIPMNPNTN DLFNAVGDGI VLCKMINLSV PDTIDERTIN KKKLTPFTIQ ENLNLALNSA SAIGCHVVNI GAEDLKEGKP YLVLGLLWQV IKIGLFADIE LSRNEALIAL LREGESLEDL MKLSPEELLL RWANYHLENA GCNKIGNFST DIKDSKAYYH LLEQVAPKGD EEGVPAVVID MSGLREKDDI QRAECMLQQA ERLGCRQFVT ATDVVRGNPK LNLAFIANLF NRYPALHKPE NQDIDWGALE GETREERTFR NWMNSLGVNP RVNHLYSDLS DALVIFQLYE KIKVPVDWNR VNKPPYPKLG GNMKKLENCN YAVELGKNQA KFSLVGIGGQ DLNEGNRTLT LALIWQLMRR YTLNILEEIG GGQKVNDDII VNWVNETLRE AKKSSSISSF KDPKISTSLP VLDLIDAIQP GSINYDLLKT ENLNDDEKLN NAKYAISMAR KIGARVYALP EDLVEVNPKM VMTVFACLMG KGMKRV
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.