missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPIAL4A Protein

A DNA sequence encoding the Homo sapiens (Human) PPIAL4A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8269-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013671

  • 8695.00 SEK / 100µg
Estimated Shipment: 09-10-2024
to see stock.

Description

Description

Specifications

Specifications

NP_001137355.1
PPIAL4A Protein
Research Use Only
PPIAL4A
Recombinant Protein
E. coli
MVNSVVFFDI TVDGKPLGRI SIKLFADKIL KTAENFRALS TGEKGFRYKG SCFHRIIPGF MCQGGDFTRH NGTGDKSIYG EKFDDENLIR KHTGSGILSM ANAGPNTNGS QFFICAAKTE WLDGKHVAFG KVKERVNIVE AMEHFGYRNS KTSKKITIAD CGQF
Greater than 90% as determined by SDS-PAGE.
653505
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers