missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPIAL4A Protein

A DNA sequence encoding the Homo sapiens (Human) PPIAL4A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8269-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023671

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001137355.1
PPIAL4A Protein
Research Use Only
PPIAL4A
Recombinant Protein
E. coli
MVNSVVFFDI TVDGKPLGRI SIKLFADKIL KTAENFRALS TGEKGFRYKG SCFHRIIPGF MCQGGDFTRH NGTGDKSIYG EKFDDENLIR KHTGSGILSM ANAGPNTNGS QFFICAAKTE WLDGKHVAFG KVKERVNIVE AMEHFGYRNS KTSKKITIAD CGQF
Greater than 90% as determined by SDS-PAGE.
653505
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers