missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP2R3B Protein
A DNA sequence encoding the Homo sapiens (Human) PPP2R3B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8271-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_037371.2 | |
PPP2R3B Protein | |
Research Use Only | |
PPP2R3B | |
Recombinant Protein | |
E. coli | |
MDLDGDGALS MFELEYFYEE QCRRLDSMAI EALPFQDCLC QMLDLVKPRT EGKITLQDLK RCKLANVFFD TFFNIEKYLD HEQKEQISLL RDGDSGGPEL SDWEKYAAEE YDILVAEETA GEPWEDGFEA ELSPVEQKLS ALRSPLAQRP FFEAPSPLGA VDLYEYACGD EDLEPL | |
Greater than 90% as determined by SDS-PAGE. |
28227 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |