missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasomal ubiquitin receptor ADRM1 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasomal ubiquitin receptor ADRM1, was expressed in the hosts and tags indicated.

2515.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_001268366.1
Gene ID (Entrez) 11047
Name Proteasomal ubiquitin receptor ADRM1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16099290
View Documents
enQuireBio™
QP7545-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16039300
View Documents
enQuireBio™
QP7545-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16089290
View Documents
enQuireBio™
QP7545-ec-100ug
100 μg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16019300
View Documents
enQuireBio™
QP7545-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16029300
View Documents
enQuireBio™
QP7545-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16009300
View Documents
enQuireBio™
QP7545-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001268366.1
Proteasomal ubiquitin receptor ADRM1 Protein
Not determined.
Full Length
Human
TTSGALFPSL VPGSRGASNK YLVEFRAGKM SLKGTTVTPD KRKGLVYIQQ TDDSLIHFCW KDRTSGNVED DLIIFPDDCE FKRVPQCPSG RVYVLKFKAG SKRLFFWMQE PKTDQDEEHC RKVNEYLNNP PMPGALGASG SSGHELSALG GEGGLQSLLG NMSHSQLMQL IGPAGLGGLG GLGALTGPGL ASLLGSSGPP GSSSSSSSRS QSAAVTPSST TSSTRATPAP SAPAAASATS PSPAPSSGNG ASTAASPTQP IQLSDLQSIL ATMNVPAGPA GGQQVDLASV LTPEIMAPIL ANADVQERLL PYLPSGESLP QTADEIQNTL TSPQFQQALG MFSAALASGQ LGPLMCQFGL PAEAVEAANK GDVEAFAKAM QNNAKPEQKE GDTKDKKDEE EDMSLD
11047
Research Use Only
ADRM1
Recombinant Protein
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.