missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein SCO2 homolog, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Protein SCO2 homolog, mitochondrial, was expressed in the hosts and tags indicated.
1640.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_001162580.1 |
---|---|
Gene ID (Entrez) | 9997 |
Name | Protein SCO2 homolog, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034830
|
enQuireBio™
QP6652-ec-10ug |
10 μg | N/A | ||||||
16074830
|
enQuireBio™
QP6652-ec-50ug |
50 μg | N/A | ||||||
16024830
|
enQuireBio™
QP6652-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16054830
|
enQuireBio™
QP6652-ec-200ug |
200 μg | N/A | ||||||
16064830
|
enQuireBio™
QP6652-ec-500ug |
500 μg | N/A | ||||||
16044830
|
enQuireBio™
QP6652-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001162580.1 | |
Protein SCO2 homolog, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9997 | |
Research Use Only | |
SCO2 | |
Recombinant Protein | |
E. coli | |
PAETGGQGQP QGPGLRTRLL ITGLFGAGLG GAWLALRAEK ERLQQQKRTE ALRQAAVGQG DFHLLDHRGR ARCKADFRGQ WVLMYFGFTH CPDICPDELE KLVQVVRQLE AEPGLPPVQP VFITVDPERD DVEAMARYVQ DFHPRLLGLT GSTKQVAQAS HSYRVYYNAG PKDEDQDYIV DHSIAIYLLN PDGLFTDYYG RSRSAEQISD SVRRHMAAFR SVLS | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title