missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human RGM-B Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 3630-RG-050
3601.63 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human RGM-B Protein is derived from NS0. The Recombinant Human RGM-B Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
13.4 kDa (N-terminus chain), 27.7 kDa (C-terminus chain) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
285704 | |
RGM-B | |
Mouse myeloma cell line,NS0-derived human RGM-B protein Gly87-Asp209 (N-terminus chain) & Pro210-Ser452 (C-terminus chain),with a C-terminal 6-His tag | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |